missing translation for 'onlineSavingsMsg'
Learn More

3-(Chloromethyl)benzoic acid, 97%, Thermo Scientific Chemicals

Product Code. 15437358
Change view
Click to view available options
Quantity:
0.5 g
10 g
Unit Size:
0.5g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15437358 10 g 10g
15427358 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15437358 Supplier Thermo Scientific Alfa Aesar Supplier No. H60854.09
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 31719-77-4 | Molecular Formula: C8H7ClO2 | Molecular Weight: 170.59 g/mol

3-(Chloromethyl)benzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Chloromethyl)benzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in methanol.

Notes
Keep container tightly sealed. Store in refrigerated conditions. Incompatible with oxidizing agents and bases.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31719-77-4
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
MDL Number MFCD00191922
InChI Key PBSUMBYSVFTMNG-UHFFFAOYSA-N
Synonym 3-chloromethyl benzoic acid, alpha-chloro-m-toluic acid, 3-chloromethyl-benzoic acid, benzoic acid, 3-chloromethyl, pubchem7446, acmc-209hog, 3-carboxybenzyl chloride, 3-chloromethyl benzoicacid, ksc497k4l, chembl67150
PubChem CID 241752
IUPAC Name 3-(chloromethyl)benzoic acid
SMILES OC(=O)C1=CC=CC(CCl)=C1

Specifications

Melting Point 136°C to 138°C
Quantity 10 g
UN Number UN3261
Solubility Information Soluble in methanol.
Formula Weight 170.59
Percent Purity 97%
Chemical Name or Material 3-(Chloromethyl)benzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.