accessibility menu, dialog, popup

UserName

CAS RN 317353-73-4

CH3HNOHNONOONOOH

CAS RN 317353-73-4

IUPAC Name: 2-(1-{4-[2-(4-{[(2-methylphenyl)carbamoyl]amino}phenyl)acetyl]morpholine-3-carbonyl}piperidin-4-yl)acetic acid
Synonyms: (1-{4-[2-(4-{[(2-methylphenyl)carbamoyl]amino}phenyl)acetyl]morpholine-3-carbonyl}piperidin-4-yl)acetic acid
Molecular Weight (g/mol): 522.6
Molecular Formula: C28H34N4O6
InChi Key: ITXAAOWFOURIHK-UHFFFAOYNA-N
SMILES: CC1=CC=CC=C1NC(=O)NC1=CC=C(CC(=O)N2CCOCC2C(=O)N2CCC(CC(O)=O)CC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results