accessibility menu, dialog, popup

UserName

CAS RN 3236-63-3

CH3OHH3COH

CAS RN 3236-63-3

IUPAC Name: 2-[(2-hydroxy-5-methylphenyl)methyl]-4-methylphenol
Synonyms: 2-[(2-hydroxy-5-methylphenyl)methyl]-4-methylphenol
Molecular Weight (g/mol): 228.29
Molecular Formula: C15H16O2
InChi Key: XZXYQEHISUMZAT-UHFFFAOYSA-N
SMILES: CC1=CC=C(O)C(CC2=CC(C)=CC=C2O)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results