missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Methylenebis(4-methylphenol) (~90%), TRC

Product Code. 30483920
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30483920 1 g 1g
30464876 500 mg 500mg
30466136 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30483920 Supplier Toronto Research Chemicals Supplier No. TRCM3041981G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3236-63-3
Molecular Formula C15 H16 O2
Molecular Weight (g/mol) 228.286
Synonym 2,2'-Methylenebis[4-methylphenol], 2,2'-Dihydroxy-5,5'-dimethyldiphenylmethane, 2,2'-Methylenebis(4-methylphenol), 2,2'-Methylenedi-p-cresol, 2-(2-Hydroxy-5-methylbenzyl)-4-methylphenol, 2-[(2-Hydroxy-5-methylphenyl)methyl]-4-methylphenol, 4,4'-Dimethyl-2,2'-methylenediphenol, Bis(2-hydroxy-5-methylphenyl)methane, Bis(4-methylphenol)-2-Methylene, M 1134
IUPAC Name 2-[(2-hydroxy-5-methylphenyl)methyl]-4-methylphenol
SMILES Cc1ccc(O)c(Cc2cc(C)ccc2O)c1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C15H16O2/c1-10-3-5-14(16)12(7-10)9-13-8-11(2)4-6-15(13)17/h3-8,16-17H,9H2,1-2H3
Formula Weight 228.115
Chemical Name or Material 2,2'-Methylenebis(4-methylphenol)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.