accessibility menu, dialog, popup

UserName

ClClClClCl

CAS RN 32598-14-4

IUPAC Name: 2,3,3',4,4'-pentachloro-1,1'-biphenyl
Synonyms: 2,3,3',4,4'-pentachloro-1,1'-biphenyl
Molecular Weight (g/mol): 326.42
Molecular Formula: C12H5Cl5
InChi Key: WIDHRBRBACOVOY-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1Cl)C1=CC=C(Cl)C(Cl)=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results