accessibility menu, dialog, popup

UserName

CAS RN 3261-87-8

IUPAC Name: 1,4-oxathiane-2,6-dione
Synonyms: 1,4-oxathiane-2,6-dione
Molecular Weight (g/mol): 132.13
Molecular Formula: C4H4O3S
InChi Key: RIIUAPMWDSRBSH-UHFFFAOYSA-N
SMILES: O=C1CSCC(=O)O1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results