missing translation for 'onlineSavingsMsg'
Learn More

Thiodiglycolic anhydride, 98%

Product Code. 11495383
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495383 25 g 25g
11485383 5 g 5g
11405393 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11495383 Supplier Thermo Scientific Alfa Aesar Supplier No. A13390.14
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3261-87-8 | C4H4O3S | 132.133 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3261-87-8
Molecular Formula C4H4O3S
Molecular Weight (g/mol) 132.133
MDL Number MFCD00051689
InChI Key RIIUAPMWDSRBSH-UHFFFAOYSA-N
Synonym thiodiglycolic anhydride, 1-oxa-4-thia-cyclohexan-2,6-dione
PubChem CID 137871
IUPAC Name 1,4-oxathiane-2,6-dione
SMILES C1C(=O)OC(=O)CS1

Specifications

Melting Point 95°C to 100°C
Quantity 25 g
Sensitivity Moisture sensitive
Beilstein 112528
Formula Weight 132.13
Percent Purity 98%
Chemical Name or Material Thiodiglycolic anhydride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.