accessibility menu, dialog, popup

UserName

BrH3CNH3CCH3CH3

CAS RN 3282-73-3

IUPAC Name: didodecyldimethylazanium bromide
Synonyms: didodecyldimethylazanium bromide
Molecular Weight (g/mol): 462.65
Molecular Formula: C26H56BrN
InChi Key: XRWMGCFJVKDVMD-UHFFFAOYSA-M
SMILES: [Br-].CCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCC
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)