missing translation for 'onlineSavingsMsg'
Learn More

(Di-n-dodecyl)dimethylammonium bromide, 98+%

Product Code. 11329789
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11329789 2 g 2g
11339789 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11329789 Supplier Thermo Scientific Alfa Aesar Supplier No. B22839.04
 Restricted Item
Estimated Shipment: 01-12-2026
Aggiungi al carrello
Aggiungi al carrello
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 3282-73-3 | C26H56BrN | 462.645 g/mol

(Di-n-dodecyl)dimethylammonium bromide is used as a phase transfer catalyst. It is also used as floor disinfectant. Further, it acts as surface-active agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Di-n-dodecyl)dimethylammonium bromide is used as a phase transfer catalyst. It is also used as floor disinfectant. Further, it acts as surface-active agent.

Solubility
Soluble in hot and cold water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 3282-73-3
Molecular Formula C26H56BrN
Molecular Weight (g/mol) 462.645
MDL Number MFCD00041969
InChI Key XRWMGCFJVKDVMD-UHFFFAOYSA-M
Synonym didodecyldimethylammonium bromide, dilauryldimethylammonium bromide, n-dodecyl-n,n-dimethyldodecan-1-aminium bromide, didodecyl dimethyl ammonium bromide, unii-7y37a25k2i, dimethyldidodecylammonium bromide, 1-dodecanaminium, n-dodecyl-n,n-dimethyl-, bromide, didodecyl dimethyl azanium bromide, 1-dodecanaminium, n-dodecyl-n,n-dimethyl-, bromide 1:1
PubChem CID 18669
IUPAC Name didodecyl(dimethyl)azanium;bromide
SMILES CCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCC.[Br-]

Specifica

Melting Point 157°C to 162°C
Linear Formula [CH3(CH2)11]2N(CH3)2Br
Quantity 2 g
Beilstein 3576662
Sensitivity Hygroscopic
Solubility Information Soluble in hot and cold water.
Formula Weight 462.65
Percent Purity ≥98%
Chemical Name or Material (Di-n-dodecyl)dimethylammonium bromide

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.