accessibility menu, dialog, popup

UserName

CAS RN 32890-89-4

CH3FHOOCl

CAS RN 32890-89-4

IUPAC Name: 2-chloro-6-fluoro-3-methylbenzoic acid
Synonyms: 2-chloro-6-fluoro-3-methylbenzoic acid
Molecular Weight (g/mol): 188.58
Molecular Formula: C8H6ClFO2
InChi Key: VCNCNVOMGXWTBZ-UHFFFAOYSA-N
SMILES: CC1=CC=C(F)C(C(O)=O)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results