accessibility menu, dialog, popup

UserName

H3COH2NFBr

CAS RN 330794-03-1

IUPAC Name: 4-bromo-5-fluoro-2-methoxyaniline
Synonyms: 4-bromo-5-fluoro-2-methoxyaniline
Molecular Weight (g/mol): 220.04
Molecular Formula: C7H7BrFNO
InChi Key: WOFOKKKWMDHLEX-UHFFFAOYSA-N
SMILES: COC1=CC(Br)=C(F)C=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results