missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-5-fluoro-2-methoxyaniline, 96%, Thermo Scientific Chemicals

Product Code. 15455698
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15455698 1 g 1g
15465698 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15455698 Supplier Thermo Scientific Alfa Aesar Supplier No. H59397.03
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
This item is not returnable. View return policy

4-Bromo-5-fluoro-2-methoxyaniline is used as a laboratory reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-5-fluoro-2-methoxyaniline is used as a laboratory reagent.

Notes
Store in a cool place. Keep the container tightly closed. Incompatible with strong-oxidizing agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 330794-03-1
Molecular Formula C7H7BrFNO
Molecular Weight (g/mol) 220.04
MDL Number MFCD21603958
InChI Key WOFOKKKWMDHLEX-UHFFFAOYSA-N
Synonym benzenamine, 4-bromo-5-fluoro-2-methoxy
PubChem CID 22347298
IUPAC Name 4-bromo-5-fluoro-2-methoxyaniline
SMILES COC1=CC(Br)=C(F)C=C1N

Specifications

Quantity 1 g
UN Number 2811
Formula Weight 220.04
Percent Purity 96%
Chemical Name or Material 4-Bromo-5-fluoro-2-methoxyaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.