CAS RN 333-47-1
IUPAC Name:
1-bromo-4-[(trifluoromethyl)sulfanyl]benzene
Synonyms:
1-bromo-4-[(trifluoromethyl)sulfanyl]benzene
Molecular Weight (g/mol):
257.07
Molecular Formula:
C7H4BrF3S
InChi Key:
CQQBVMXLTBXNLB-UHFFFAOYSA-N
SMILES:
FC(F)(F)SC1=CC=C(Br)C=C1
1
–
1
of
1
results
1
1-Bromo-4-(trifluoromethylthio)benzene, 97%
CAS: 333-47-1 Molecular Formula: C7H4BrF3S Molecular Weight (g/mol): 257.068 MDL Number: MFCD00040840 InChI Key: CQQBVMXLTBXNLB-UHFFFAOYSA-N Synonym: 4-trifluoromethylthio bromobenzene,1-bromo-4-trifluoromethyl sulfanyl benzene,4-bromophenyl trifluoromethyl sulfide,4-bromophenyl trifluoromethyl sulphide,1-bromo-4-trifluoromethylthio benzene,1-bromo-4-trifluoromethylthiobenzene,4-bromophenyl trifluoromethyl sulfane,1-bromo-4-trifluoromethyl thio benzene,4-bromo-1-trifluoromethylthio benzene,1-bromo-4-trifluoromethylsulfanyl benzene PubChem CID: 736413 IUPAC Name: 1-bromo-4-(trifluoromethylsulfanyl)benzene SMILES: C1=CC(=CC=C1SC(F)(F)F)Br