missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-(trifluoromethylthio)benzene, 97%

Product Code. 11469896
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11469896 5 g 5g
11459896 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11469896 Supplier Thermo Scientific Alfa Aesar Supplier No. B20874.06
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 333-47-1 | C7H4BrF3S | 257.068 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 333-47-1
Molecular Formula C7H4BrF3S
Molecular Weight (g/mol) 257.068
MDL Number MFCD00040840
InChI Key CQQBVMXLTBXNLB-UHFFFAOYSA-N
Synonym 4-trifluoromethylthio bromobenzene, 1-bromo-4-trifluoromethyl sulfanyl benzene, 4-bromophenyl trifluoromethyl sulfide, 4-bromophenyl trifluoromethyl sulphide, 1-bromo-4-trifluoromethylthio benzene, 1-bromo-4-trifluoromethylthiobenzene, 4-bromophenyl trifluoromethyl sulfane, 1-bromo-4-trifluoromethyl thio benzene, 4-bromo-1-trifluoromethylthio benzene, 1-bromo-4-trifluoromethylsulfanyl benzene
PubChem CID 736413
IUPAC Name 1-bromo-4-(trifluoromethylsulfanyl)benzene
SMILES C1=CC(=CC=C1SC(F)(F)F)Br

Specifications

Density 1.71
Boiling Point 199°C to 200°C
Flash Point 83°C (181°F)
Refractive Index 1.517
Quantity 5 g
Beilstein 2361926
Formula Weight 257.07
Percent Purity 97%
Chemical Name or Material 1-Bromo-4-(trifluoromethylthio)benzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.