accessibility menu, dialog, popup

UserName

CH3OOBrF

CAS RN 334792-52-8

IUPAC Name: methyl 3-bromo-5-fluorobenzoate
Synonyms: methyl 3-bromo-5-fluorobenzoate
Molecular Weight (g/mol): 233.04
Molecular Formula: C8H6BrFO2
InChi Key: JERAACCIOWRRQA-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(F)=CC(Br)=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results