missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-bromo-5-fluorobenzoate, Thermo Scientific™

Product Code. 10649000
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10649000 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10649000 Supplier Thermo Scientific Maybridge Supplier No. MAY00092DA
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 334792-52-8
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C8H6BrFO2
MDL Number MFCD07780735
Quantity 1 g
Synonym 3-bromo-5-fluorobenzoic acid methyl ester, benzoic acid, 3-bromo-5-fluoro-, methyl ester, acmc-209i1c, methyl 5-bromo-3-fluorobenzoate, methyl-3-bromo-5-fluorobenzoate, 3-bromo-5-fluoro-benzoic acid methyl ester, benzoicacid, 3-bromo-5-fluoro-, methyl ester
InChI Key JERAACCIOWRRQA-UHFFFAOYSA-N
SMILES COC(=O)C1=CC(F)=CC(Br)=C1
IUPAC Name methyl 3-bromo-5-fluorobenzoate
Molecular Weight (g/mol) 233.04
PubChem CID 21942598
Formula Weight 233.04
Percent Purity 97%
Chemical Name or Material Methyl 3-bromo-5-fluorobenzoate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.