accessibility menu, dialog, popup

UserName

HOOFFFFFFFFFFFFFFF

CAS RN 335-67-1

IUPAC Name: pentadecafluorooctanoic acid
Synonyms: perfluorooctanoic acid perfluorooctanoic acid, cobalt (2+) salt cobalt (2+) salt perfluorooctanoic acid monosilver (1+) salt perfluorooctanoic acid perfluorooctanoic acid, chromium (3+) salt pfoa cpd monosodium salt perfluorooctanoic acid cesium salt perfluorooctanoic acid lithium salt perfluorooctanoic acid anhydride perfluorooctanoic acid
Molecular Weight (g/mol): 414.07
Molecular Formula: C8HF15O2
InChi Key: SNGREZUHAYWORS-UHFFFAOYSA-N
SMILES: OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (3)
Boiling Point 
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)