missing translation for 'onlineSavingsMsg'
Learn More

Perfluorooctanoic Acid, TRC

Product Code. 30133340
Change view
Click to view available options
Quantity:
2.5 g
25 g
Unit Size:
2.5g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133340 25 g 25g
30011237 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133340 Supplier Toronto Research Chemicals Supplier No. TRCP28650025G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 335-67-1
Molecular Formula C8 H F15 O2
Molecular Weight (g/mol) 414.07
Synonym Octanoic acid, pentadecafluoro- (8CI,9CI), 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Pentadecafluorooctanoic acid, A 5717, EF 201, Eftop EF 201, NSC 95114, PFOA, Pentadecafluoro-n-octanoic acid, Pentadecafluorooctanoic acid, Perfluoro-1-heptanecarboxylic acid, Perfluorocaprylic acid, Perfluoroheptanecarboxylic acid, Perfluorooctanoic acid, n-Perfluorooctanoic acid, Perfluorooctanoic Acid, Octanoic acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro-
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctanoic acid
SMILES OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Quantity 25 g
InChI Formula InChI=1S/C8HF15O2/c9-2(10,1(24)25)3(11,12)4(13,14)5(15,16)6(17,18)7(19,20)8(21,22)23/h(H,24,25)
Formula Weight 413.9737
Percent Purity >95%
Chemical Name or Material Perfluorooctanoic acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.