accessibility menu, dialog, popup

UserName

FFFFFFFOOOFFFFFFF

CAS RN 336-59-4

IUPAC Name: 2,2,3,3,4,4,4-heptafluorobutanoyl 2,2,3,3,4,4,4-heptafluorobutanoate
Synonyms: heptafluorobutyric anhydride HFBA
Molecular Weight (g/mol): 410.06
Molecular Formula: C8F14O3
InChi Key: UFFSXJKVKBQEHC-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C(F)(F)C(=O)OC(=O)C(F)(F)C(F)(F)C(F)(F)F
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)