missing translation for 'onlineSavingsMsg'
Learn More

Heptafluorobutyric anhydride, 98%

Product Code. 11499622
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499622 100 g 100g
11479622 5 g 5g
11489622 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499622 Supplier Thermo Scientific Alfa Aesar Supplier No. A10744.22
 Restricted Item
Estimated Shipment: 10-11-2026
Læg i indkøbskurv
Læg i indkøbskurv
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

CAS: 336-59-4 | C8F14O3 | 410.06 g/mol

Heptafluorobutyric anhydride is used as a derivatization reagent for gas chromatography - mass mass spectrometry analysis of the stereoisomer content of synthetic peptides. It is a GC derivatizing agent used to prepare volatile derivatives of amino acids and other biologically active compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Heptafluorobutyric anhydride is used as a derivatization reagent for gas chromatography - mass mass spectrometry analysis of the stereoisomer content of synthetic peptides. It is a GC derivatizing agent used to prepare volatile derivatives of amino acids and other biologically active compounds.

Solubility
Miscible with halogenated solvents. Immiscible with polar solvents.

Notes
Incompatible with moist air, water, strong oxidizing agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 336-59-4
Molecular Formula C8F14O3
Molecular Weight (g/mol) 410.06
MDL Number MFCD00000432
InChI Key UFFSXJKVKBQEHC-UHFFFAOYSA-N
Synonym heptafluorobutyric anhydride, perfluorobutyric anhydride, hfba, heptafluorobutanoic anhydride, 2,2,3,3,4,4,4-heptafluorobutanoic anhydride, butanoic acid, heptafluoro-, anhydride, heptafluoro-n-butyric anhydride, heptafluorobutyric acid anhydride, butanoic acid, 2,2,3,3,4,4,4-heptafluoro-, 1,1'-anhydride, heptafluorobutyric anhydride, for gc derivatization
PubChem CID 67643
ChEBI CHEBI:39424
IUPAC Name 2,2,3,3,4,4,4-heptafluorobutanoyl 2,2,3,3,4,4,4-heptafluorobutanoate
SMILES FC(F)(F)C(F)(F)C(F)(F)C(=O)OC(=O)C(F)(F)C(F)(F)C(F)(F)F

Tekniske data

Melting Point -43°C
Density 1.662
Boiling Point 108°C to 110°C
Odor Stench
Linear Formula (CF3CF2CF2CO)2O
Refractive Index 1.287
Quantity 100 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Miscible with halogenated solvents. Immiscible with polar solvents.
Formula Weight 410.06
Percent Purity 98%
Chemical Name or Material Heptafluorobutyric anhydride
Vis mere Vis mindre
compliance-icons
Produktidentifikator
  • Heptafluorobutyric anhydride
Signalord
  • Danger
Farekategori
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Faresætning
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Sikkerhedssætning
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplerende oplysninger
  • MIXTURE LIST-Contain: Heptafluorobutyric anhydride

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.