missing translation for 'onlineSavingsMsg'
Learn More

Heptafluorobutyric anhydride, 98%

Product Code. 10522961 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10522961 25 g Glass bottle 25g
10144090 5 g Glass bottle 5g
10309143 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10522961 Supplier Thermo Scientific Acros Supplier No. 169260250
  • ONLINE EXCLUSIVE PRICE
    190.00 €
    175.00 € / 25g
    Save 15.00 €
    7% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 336-59-4 | C8F14O3 | 410.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Acylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 336-59-4
Molecular Formula C8F14O3
Molecular Weight (g/mol) 410.06
MDL Number MFCD00000432
InChI Key UFFSXJKVKBQEHC-UHFFFAOYSA-N
Synonym heptafluorobutyric anhydride, perfluorobutyric anhydride, hfba, heptafluorobutanoic anhydride, 2,2,3,3,4,4,4-heptafluorobutanoic anhydride, butanoic acid, heptafluoro-, anhydride, heptafluoro-n-butyric anhydride, heptafluorobutyric acid anhydride, butanoic acid, 2,2,3,3,4,4,4-heptafluoro-, 1,1'-anhydride, heptafluorobutyric anhydride, for gc derivatization
PubChem CID 67643
ChEBI CHEBI:39424
IUPAC Name 2,2,3,3,4,4,4-heptafluorobutanoyl 2,2,3,3,4,4,4-heptafluorobutanoate
SMILES FC(F)(F)C(F)(F)C(F)(F)C(=O)OC(=O)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Melting Point -43°C
Density 1.6500g/mL
Boiling Point 108°C to 111°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula (CF3CF2CF2CO)2O
Quantity 25 g
Specific Gravity 1.65
Solubility Information Solubility in water: reacts with water forming heptafluorobutyric acid
Formula Weight 410.05
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Heptafluorobutyric anhydride
Show More Show Less
compliance-icons
Product Identifier
  • Heptafluorobutyric anhydride
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.