accessibility menu, dialog, popup

UserName

CAS RN 3374-22-9

IUPAC Name: 2-amino-3-sulfanylpropanoic acid
Synonyms: cysteine zystein Hcys cisteina
Molecular Weight (g/mol): 121.15
Molecular Formula: C3H7NO2S
InChi Key: XUJNEKJLAYXESH-UHFFFAOYNA-N
SMILES: NC(CS)C(O)=O
Molecular Weight (g/mol) 
  • (5)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)