missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Cysteine, 97%

Product Code. 11330686 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass Bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11330686 50 g Glass Bottle 50g
11320686 10 g Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11330686 Supplier Thermo Scientific Chemicals Supplier No. 230120500
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3374-22-9 | C3H7NO2S | 121.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3374-22-9
Molecular Formula C3H7NO2S
Molecular Weight (g/mol) 121.15
MDL Number MFCD00004881
InChI Key XUJNEKJLAYXESH-UHFFFAOYNA-N
Synonym dl-cysteine, cysteine, 2-amino-3-mercaptopropanoic acid, dl-cystein, cysteine, dl, +--cysteine, dl-cysteina polish, h-dl-cys-oh, 2-amino-3-mercaptopropionic acid, l-hsch2ch nh2 cooh
PubChem CID 594
ChEBI CHEBI:15356
IUPAC Name 2-amino-3-sulfanylpropanoic acid
SMILES NC(CS)C(O)=O

Specifications

Melting Point 225.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Linear Formula HSCH2CH(NH2)CO2H
Beilstein 04, IV, 3145
Merck Index 10, 2775
Solubility Information Solubility in water: soluble. Other solubilities: freely soluble in alcohol, insolublein ether, acetone, benzene, carbon, disulfide, ethyl acetate and carbon tetrachloride
Quantity 50 g
Formula Weight 121.15
Percent Purity 97%
Physical Form Powder
Chemical Name or Material DL-Cysteine, 99%
Show More Show Less
compliance-icons
Product Identifier
  • DL-Cysteine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.