accessibility menu, dialog, popup

UserName

H3COHCH2

CAS RN 3391-86-4

IUPAC Name: oct-1-en-3-ol
Synonyms: 1-octen-3-ol mushroom alcohol epa pesticide chemical code 069037 1-octen-3-ol (natural) (R)-1-octen-3-ol 1-octen-3-ol, (R)-isomer (+-)-1-octen-3-ol 1-octen-3-ol, (+-)- 1-octen-3-ol, (+-)-isomer (r)-isomer 1-octen-3-ol
Molecular Weight (g/mol): 128.22
Molecular Formula: C8H16O
InChi Key: VSMOENVRRABVKN-UHFFFAOYNA-N
SMILES: CCCCCC(O)C=C
Molecular Weight (g/mol) 
  • (2)
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)