missing translation for 'onlineSavingsMsg'
Learn More

1-Octen-3-ol, 98%

Product Code. 10637672 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10637672 25 g Glass bottle 25g
10406242 5 g Glass bottle 5g
10616524 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10637672 Supplier Thermo Scientific Acros Supplier No. 129470250
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3391-86-4 | C8H16O | 128.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3391-86-4
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.22
MDL Number MFCD00004589
InChI Key VSMOENVRRABVKN-UHFFFAOYSA-N
Synonym 1-octen-3-ol, vinyl amyl carbinol, 1-vinylhexanol, 3-hydroxy-1-octene, amyl vinyl carbinol, mushroom alcohol, matsutake alcohol, vinyl hexanol, pentyl vinyl carbinol, matsuica alcohol
PubChem CID 18827
ChEBI CHEBI:34118
IUPAC Name oct-1-en-3-ol
SMILES CCCCCC(C=C)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8370g/mL
Boiling Point 174.0°C
Flash Point 68°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)4CH(OH)CH=CH2
Refractive Index 1.4340 to 1.4420
Quantity 25 g
Specific Gravity 0.837
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in ethanol
Formula Weight 128.22
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Octen-3-ol
Show More Show Less
compliance-icons
Product Identifier
  • 1-Octen-3-ol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.