missing translation for 'onlineSavingsMsg'
Learn More

(±)-1-Octen-3-ol, TRC

Product Code. 30133266
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133266 25 g 25g
30011088 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133266 Supplier Toronto Research Chemicals Supplier No. TRCO23920525G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3391-86-4
Molecular Formula C8 H16 O
Molecular Weight (g/mol) 128.21
Synonym 1-Octen-3-ol, (±)-1-Octen-3-ol, 1-Vinylhexanol, 3-Hydroxy-1-octene, Amyl vinyl carbinol, Moguchun, Morillol, Morrilol, NSC 87563, Pentylvinylcarbinol, Primaverol, Vinyl amyl carbinol, dl-1-Octen-3-ol, n-Oct-1-en-3-ol
IUPAC Name oct-1-en-3-ol
SMILES CCCCCC(O)C=C

Specifications

Quantity 25 g
InChI Formula InChI=1S/C8H16O/c1-3-5-6-7-8(9)4-2/h4,8-9H,2-3,5-7H2,1H3
Formula Weight 128.1201
Chemical Name or Material 1-Octen-3-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.