accessibility menu, dialog, popup

UserName

BNSOOHOHO

CAS RN 342404-46-0

IUPAC Name: [1-(benzenesulfonyl)-1H-indol-2-yl]boronic acid
Synonyms: 1-(benzenesulfonyl)indol-2-ylboronic acid
Molecular Weight (g/mol): 301.12
Molecular Formula: C14H12BNO4S
InChi Key: HXWLCYMHOULBJZ-UHFFFAOYSA-N
SMILES: OB(O)C1=CC2=CC=CC=C2N1S(=O)(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results