missing translation for 'onlineSavingsMsg'
Learn More

1-(Phenylsulfonyl)-1H-indol-2-ylboronic acid, 97%, Thermo Scientific™

Product Code. 10712377
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10712377 5 g Amber Glass Bottle 5g
10477992 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10712377 Supplier Thermo Scientific Maybridge Supplier No. CC03312DE
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 342404-46-0
Molecular Formula C14H12BNO4S
Molecular Weight (g/mol) 301.12
MDL Number MFCD03086094
InChI Key HXWLCYMHOULBJZ-UHFFFAOYSA-N
Synonym 1-phenylsulfonyl-2-indoleboronic acid, 1-phenylsulfonyl-1h-indol-2-yl boronic acid, 1-phenylsulfonyl-1h-indol-2-ylboronic acid, 1-phenylsulfonyl-2-indolylboronic acid, 1-phenylsulfonyl indole-2-boronic acid, 1-benzenesulfonyl indol-2-ylboronic acid, boronic acid,b-1-phenylsulfonyl-1h-indol-2-yl, 1-benzenesulfonyl indol-2-yl boronic acid, acmc-209i5v, 1-phenylsulfonylindole-2-boronic acid
PubChem CID 2776228
IUPAC Name [1-(benzenesulfonyl)indol-2-yl]boronic acid
SMILES OB(O)C1=CC2=CC=CC=C2N1S(=O)(=O)C1=CC=CC=C1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 301.13
Chemical Name or Material 1-(Phenylsulfonyl)-1H-indol-2-ylboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.