accessibility menu, dialog, popup

UserName

CAS RN 34252-44-3

H3CNNOOH

CAS RN 34252-44-3

IUPAC Name: 2-methyl-2H-indazole-3-carboxylic acid
Synonyms: 2-methylindazole-3-carboxylic acid
Molecular Weight (g/mol): 176.18
Molecular Formula: C9H8N2O2
InChi Key: ABRISPFTOZIXMP-UHFFFAOYSA-N
SMILES: CN1N=C2C=CC=CC2=C1C(O)=O
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results