accessibility menu, dialog, popup

UserName

CAS RN 3435-51-6

IUPAC Name: 4-ethenylbenzonitrile
Synonyms: 4-ethenylbenzonitrile
Molecular Weight (g/mol): 129.16
Molecular Formula: C9H7N
InChi Key: SNTUCKQYWGHZPK-UHFFFAOYSA-N
SMILES: C=CC1=CC=C(C=C1)C#N
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)