missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanostyrene, 95%, stabilized

Product Code. 11915621 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11915621 10 g Glass bottle 10g
10104690 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11915621 Supplier Thermo Scientific Acros Supplier No. 292910100
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3435-51-6 | C9H7N | 129.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3435-51-6
Molecular Formula C9H7N
Molecular Weight (g/mol) 129.16
MDL Number MFCD00080445
InChI Key SNTUCKQYWGHZPK-UHFFFAOYSA-N
PubChem CID 76967
IUPAC Name 4-ethenylbenzonitrile
SMILES C=CC1=CC=C(C=C1)C#N

Specifications

Color Colorless to Yellow
Boiling Point 56.0°C to 58.0°C (10.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.5765 to 1.5785
Quantity 10 g
Synonym 4-cyanostyrene, 4-vinylbenzonitrile, benzonitrile, 4-ethenyl, p-cyanostyrene, 4-vinyl-benzonitrile, benzonitrile, p-vinyl, p-vinylbenzonitrile, acmc-20amk5, benzonitrile,4-ethenyl, 4-vinylbenzenecarbonitrile
Solubility Information Solubility in water: insoluble
Stabilizer 0.05% tert.butylcatechol
Formula Weight 129.16
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 4-Cyanostyrene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.