CAS RN 347909-09-5
IUPAC Name:
1-(2-fluorobenzoyl)pyrrolidine
Synonyms:
1-(2-fluorobenzoyl)pyrrolidine
Molecular Weight (g/mol):
193.22
Molecular Formula:
C11H12FNO
InChi Key:
AZBXHPRYPHBYCV-UHFFFAOYSA-N
SMILES:
FC1=CC=CC=C1C(=O)N1CCCC1
1
–
1
of
1
results
1
1-(2-Fluorobenzoyl)pyrrolidine, 97%, Thermo Scientific™
CAS: 347909-09-5 Molecular Formula: C11H12FNO Molecular Weight (g/mol): 193.221 MDL Number: MFCD00460952 InChI Key: AZBXHPRYPHBYCV-UHFFFAOYSA-N Synonym: 1-2-fluorobenzoyl pyrrolidine,2-fluorophenyl 1-pyrrolidinyl methanone,2-fluorophenyl-pyrrolidin-1-ylmethanone PubChem CID: 3497148 IUPAC Name: (2-fluorophenyl)-pyrrolidin-1-ylmethanone SMILES: C1CCN(C1)C(=O)C2=CC=CC=C2F