accessibility menu, dialog, popup

UserName

CAS RN 348622-88-8

OHNOHNNOONH2

CAS RN 348622-88-8

IUPAC Name: 5-[5-cyclohexyl-1-(hydroxycarbamoyl)pentan-2-yl]-1,2,4-oxadiazole-3-carboxamide
Synonyms: 5-[5-cyclohexyl-1-(hydroxycarbamoyl)pentan-2-yl]-1,2,4-oxadiazole-3-carboxamide
Molecular Weight (g/mol): 324.38
Molecular Formula: C15H24N4O4
InChi Key: ARJCBSRIPGJMAD-UHFFFAOYNA-N
SMILES: NC(=O)C1=NOC(=N1)C(CCCC1CCCCC1)CC(=O)NO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results