accessibility menu, dialog, popup

UserName

CH

CAS RN 34993-56-1

IUPAC Name: 1-ethynylpyrene
Synonyms: 1-ethynylpyrene
Molecular Weight (g/mol): 226.28
Molecular Formula: C18H10
InChi Key: VEBUBSLYGRMOSZ-UHFFFAOYSA-N
SMILES: C#CC1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results