missing translation for 'onlineSavingsMsg'
Learn More

1-Ethynylpyrene, 96%

Product Code. 11393996
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11393996 5 g 5g
11383996 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11393996 Supplier Thermo Scientific Alfa Aesar Supplier No. L17227.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 34993-56-1 | C18H10 | 226.28 g/mol

1-Ethynylpyrene is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Ethynylpyrene is used as a pharmaceutical intermediate.

Solubility
Soluble in chloroform, dichloromethane and toluene. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 34993-56-1
Molecular Formula C18H10
Molecular Weight (g/mol) 226.28
MDL Number MFCD02093933
InChI Key VEBUBSLYGRMOSZ-UHFFFAOYSA-N
Synonym 1-ethynyl pyrene, pyrene, 1-ethynyl, ethynylpyrene, 1-pyrenylethyne, ccris 4256, acmc-20aov2
PubChem CID 154905
IUPAC Name 1-ethynylpyrene
SMILES C#CC1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34

Spezifikation

Melting Point 113°C to 116°C
Quantity 5 g
Beilstein 2365008
Solubility Information Soluble in chloroform,dichloromethane and toluene. Insoluble in water.
Formula Weight 226.28
Percent Purity 96%
Chemical Name or Material 1-Ethynylpyrene

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.