accessibility menu, dialog, popup

UserName

CAS RN 352-32-9

CH3F

CAS RN 352-32-9

IUPAC Name: 1-fluoro-4-methylbenzene
Synonyms: P-fluorotoluene
Molecular Weight (g/mol): 110.13
Molecular Formula: C7H7F
InChi Key: WRWPPGUCZBJXKX-UHFFFAOYSA-N
SMILES: CC1=CC=C(F)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)