missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorotoluene, 99%

Product Code. 11414173
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11414173

Brand: Thermo Scientific Alfa Aesar A10365.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 352-32-9 | C7H7F | 110.131 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 352-32-9
Molecular Formula C7H7F
Molecular Weight (g/mol) 110.131
MDL Number MFCD00000358
InChI Key WRWPPGUCZBJXKX-UHFFFAOYSA-N
Synonym 4-fluorotoluene, p-fluorotoluene, benzene, 1-fluoro-4-methyl, p-fluoromethylbenzene, toluene, p-fluoro, 1-methyl-4-fluorobenzene, p-fluoromethyl benzene, unii-nud6wz1b8f, p-tolyl fluoride, para-fluorotoluene
PubChem CID 9603
IUPAC Name 1-fluoro-4-methylbenzene
SMILES CC1=CC=C(C=C1)F

Specifications

Melting Point -56°C
Density 1.001
Boiling Point 115°C to 116°C
Flash Point 10°C (50°F)
Odor Aromatic
Refractive Index 1.4685
Quantity 500 g
UN Number UN2388
Beilstein 1362373
Merck Index 14,4180
Formula Weight 110.13
Percent Purity 99%
Chemical Name or Material 4-Fluorotoluene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.