accessibility menu, dialog, popup

UserName

CAS RN 35444-44-1

H3COOClO

CAS RN 35444-44-1

IUPAC Name: methyl 6-chloro-6-oxohexanoate
Synonyms: methyl 6-chloro-6-oxohexanoate
Molecular Weight (g/mol): 178.61
Molecular Formula: C7H11ClO3
InChi Key: HDLGIEZOMYJKAK-UHFFFAOYSA-N
SMILES: COC(=O)CCCCC(Cl)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)