CAS RN 3563-27-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
17Beta-Dihydro Equilin, TRC
CAS: 3563-27-7 Molecular Formula: C18 H22 O2 Molecular Weight (g/mol): 270.37 Synonym: Estra-1,3,5(10),7-tetraene-3,17-diol, (17β)-,Estra-1,3,5(10),7-tetraene-3,17β-diol (6CI,7CI,8CI),(17β)-Estra-1,3,5(10),7-tetraene-3,17-diol,17β-Dihydroequilin,7-Dehydroestradiol,NSC 12170,β-Dihydroequilin IUPAC Name: (9S,13S,14S,17S)-13-methyl-6,9,11,12,14,15,16,17-octahydrocyclopenta[a]phenanthrene-3,17-diol SMILES: C[C@]12CC[C@H]3C(=CCc4cc(O)ccc34)[C@@H]1CC[C@@H]2O