accessibility menu, dialog, popup

UserName

CH3ONHN

CAS RN 35704-19-9

IUPAC Name: N-(4-cyanophenyl)acetamide
Synonyms: acetamide, N-(4-cyanophenyl)-
Molecular Weight (g/mol): 160.18
Molecular Formula: C9H8N2O
InChi Key: UFKRTEWFEYWIHD-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(C=C1)C#N
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)