missing translation for 'onlineSavingsMsg'
Learn More

4'-Cyanoacetanilide, 98%, Thermo Scientific™

Product Code. 10766423 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Packungsgröße:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Packaging Packungsgröße
10766423 5 g Glass bottle 5g
10565833 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Product Code. 10766423 Supplier Thermo Scientific Acros Supplier No. 373250050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 35704-19-9
Molecular Formula C9H8N2O
Molecular Weight (g/mol) 160.18
MDL Number MFCD00001814
InChI Key UFKRTEWFEYWIHD-UHFFFAOYSA-N
Synonym n-4-cyanophenyl acetamide, 4-cyanoacetanilide, 4-acetamidobenzonitrile, acetamide, n-4-cyanophenyl, 4'-cyanoacetanilide, p-acetamidobenzonitirle, acetanilide, 4'-cyano, p-acetylamino-benzonitrile, p-cyanoacetanilide, p-acetaminobenzonitrile
PubChem CID 37256
IUPAC Name N-(4-cyanophenyl)acetamide
SMILES CC(=O)NC1=CC=C(C=C1)C#N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 204°C to 208°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CONHC6H4CN
Quantity 5 g
Formula Weight 160.18
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 4'-Cyanoacetanilide
Show More Show Less
compliance-icons
Product Identifier
  • 4'-Cyanoacetanilide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.