accessibility menu, dialog, popup

UserName

FFFONHFFF

CAS RN 35853-41-9

IUPAC Name: 2,8-bis(trifluoromethyl)-1,4-dihydroquinolin-4-one
Synonyms: 2,8-bis(trifluoromethyl)-1H-quinolin-4-one
Molecular Weight (g/mol): 281.16
Molecular Formula: C11H5F6NO
InChi Key: JIWHKBAFGFPZKM-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(=O)C2=CC=CC(=C2N1)C(F)(F)F
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results