accessibility menu, dialog, popup

UserName

NNOOHNH2

CAS RN 361442-04-8

IUPAC Name: 2-[2-amino-2-(3-hydroxyadamantan-1-yl)acetyl]-2-azabicyclo[3.1.0]hexane-3-carbonitrile
Synonyms: saxagliptin
Molecular Weight (g/mol): 315.42
Molecular Formula: C18H25N3O2
InChi Key: QGJUIPDUBHWZPV-UHFFFAOYNA-N
SMILES: NC(C(=O)N1C2CC2CC1C#N)C12CC3CC(CC(O)(C3)C1)C2
Molecular Weight (g/mol) 
  • (7)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (7)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)