accessibility menu, dialog, popup

UserName

CAS RN 363-52-0

OHFHO

CAS RN 363-52-0

IUPAC Name: 3-fluorobenzene-1,2-diol
Synonyms: 3-fluorocatechol
Molecular Weight (g/mol): 128.1
Molecular Formula: C6H5FO2
InChi Key: DXOSJQLIRGXWCF-UHFFFAOYSA-N
SMILES: OC1=CC=CC(F)=C1O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)