missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorocatechol, TRC

Product Code. 30137362
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137362 50 g 50g
30008712 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137362 Supplier Toronto Research Chemicals Supplier No. TRCF58882550G
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 363-52-0
Molecular Formula C6 H5 F O2
Molecular Weight (g/mol) 128.101
Synonym 3-Fluoro-1,2-benzenediol (ACI), Pyrocatechol, 3-fluoro- (8CI), 1,2-Dihydroxy-3-fluorobenzene, 3-Fluoro-2-hydroxyphenol, 3-Fluorocatechol
IUPAC Name 3-fluorobenzene-1,2-diol
SMILES Oc1cccc(F)c1O

Specifications

Quantity 50 g
InChI Formula InChI=1S/C6H5FO2/c7-4-2-1-3-5(8)6(4)9/h1-3,8-9H
Formula Weight 128.027
Chemical Name or Material 3-Fluorocatechol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.