CAS RN 3650-09-7
CAS RN 3650-09-7
IUPAC Name:
(4aR,10aS)-5,6-dihydroxy-1,1-dimethyl-7-(propan-2-yl)-1,2,3,4,4a,9,10,10a-octahydrophenanthrene-4a-carboxylic acid
Synonyms:
salvin carnosic acid
Molecular Weight (g/mol):
332.44
Molecular Formula:
C20H28O4
InChi Key:
QRYRORQUOLYVBU-VBKZILBWSA-N
SMILES:
CC(C)C1=CC2=C(C(O)=C1O)[C@]1(CCCC(C)(C)[C@@H]1CC2)C(O)=O
1
–
1
of
1
results
1
Carnosic acid, MedChemExpress
MedChemExpress Carnosic acid has demonstrated inhibition of oxidative stress and inflammation, suppression of cell proliferation, and antibacterial activity.