CAS RN 3663-79-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Methyl 1,4-Benzodioxan-2-carboxylate, TRC
CAS: 3663-79-4 Molecular Formula: C10 H10 O4 Molecular Weight (g/mol): 194.18 Synonym: 1,4-Benzodioxin-2-carboxylic acid, 2,3-dihydro-, methyl ester,1,4-Benzodioxan-2-carboxylic acid, methyl ester (6CI,7CI,8CI),Methyl 1,4-benzodioxan-2-carboxylate,Methyl 1,4-benzodioxane-2-carboxylate,Methyl (2RS)-1,4-benzodioxane-2-carboxylate IUPAC Name: methyl 2,3-dihydro-1,4-benzodioxine-3-carboxylate SMILES: COC(=O)C1COc2ccccc2O1
2
Methyl (2RS)-1,4-Benzodioxane-2-carboxylate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.