CAS RN 368-77-4
IUPAC Name:
3-(trifluoromethyl)benzonitrile
Synonyms:
A,A,A-trifluor-M-tolunitril
Molecular Weight (g/mol):
171.12
Molecular Formula:
C8H4F3N
InChi Key:
OGOBINRVCUWLGN-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=CC=CC(=C1)C#N
1
–
1
of
1
results
1
3-(Trifluoromethyl)benzonitrile, 98%
CAS: 368-77-4 Molecular Formula: C8H4F3N Molecular Weight (g/mol): 171.122 MDL Number: MFCD00001807 InChI Key: OGOBINRVCUWLGN-UHFFFAOYSA-N Synonym: 3-trifluoromethyl benzonitrile,3-cyanobenzotrifluoride,m-trifluoromethylbenzonitrile,alpha,alpha,alpha-trifluoro-m-tolunitrile,benzonitrile, 3-trifluoromethyl,a,a,a-trifluoro-m-tolunitrile,unii-y7zd7dk3ay,alpha,alpha,alpha-trifluoro-m-toluonitrile,y7zd7dk3ay,a,a,a-trifluor-m-tolunitril PubChem CID: 67783 IUPAC Name: 3-(trifluoromethyl)benzonitrile SMILES: C1=CC(=CC(=C1)C(F)(F)F)C#N