missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)benzonitrile, 98%

Product Code. 11458886
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11458886 100 g 100g
11319549 5 g 5g
11448886 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11458886 Supplier Thermo Scientific Alfa Aesar Supplier No. A17029.22
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 368-77-4 | C8H4F3N | 171.122 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 368-77-4
Molecular Formula C8H4F3N
Molecular Weight (g/mol) 171.122
MDL Number MFCD00001807
InChI Key OGOBINRVCUWLGN-UHFFFAOYSA-N
Synonym 3-trifluoromethyl benzonitrile, 3-cyanobenzotrifluoride, m-trifluoromethylbenzonitrile, alpha,alpha,alpha-trifluoro-m-tolunitrile, benzonitrile, 3-trifluoromethyl, a,a,a-trifluoro-m-tolunitrile, unii-y7zd7dk3ay, alpha,alpha,alpha-trifluoro-m-toluonitrile, y7zd7dk3ay, a,a,a-trifluor-m-tolunitril
PubChem CID 67783
IUPAC Name 3-(trifluoromethyl)benzonitrile
SMILES C1=CC(=CC(=C1)C(F)(F)F)C#N

Specifications

Melting Point 14°C to 18°C
Density 1.28
Boiling Point 188°C to 190°C
Flash Point 72°C (161°F)
Refractive Index 1.459
Quantity 100 g
UN Number UN3276
Beilstein 1868102
Formula Weight 171.12
Percent Purity 98%
Chemical Name or Material 3-(Trifluoromethyl)benzonitrile
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.