CAS RN 3687-22-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Meptyldinocap-phenol 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
2,4-Dinitro-6-(1-methylheptyl)phenol, TRC
CAS: 3687-22-7 Molecular Formula: C14 H20 N2 O5 Molecular Weight (g/mol): 296.32 Synonym: 2-(1-Methylheptyl)-4,6-dinitrophenol,2,4-DNOP,2,4-Dinitro-6-(1-methylheptyl)phenol,2,4-Dinitro-6-(1'-methylheptyl)phenol,2-(2-Hydroxy-3,5-dinitrophenyl)octane,DNOP,Meptyldinocap-phenol IUPAC Name: 2,4-dinitro-6-octan-2-ylphenol SMILES: CCCCCCC(C)c1cc(cc(c1O)[N+](=O)[O-])[N+](=O)[O-]